missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TXNDC5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583241
This item is not returnable.
View return policy
Description
Immunogen sequence: TQHYELCSGN QVRGYPTLLW FRDGKKVDQY KGKRDLESLR EYVESQLQRT ETGATETVTP SEAPVLAAEP EADKGTVLAL TENNFD Highest antigen sequence indentity to the following orthologs: Rat - 80%, Mouse - 79%.
This gene encodes a protein-disulfide isomerase. Its expression is induced by hypoxia and its role may be to protect hypoxic cells from apoptosis. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
| TXNDC5 | |
| Polyclonal | |
| Unconjugated | |
| Txndc5 | |
| AL022641; ENDOPDI; endoplasmic reticulum protein ERp46; endoplasmic reticulum resident protein 46; endothelial protein disulphide isomerase; ER protein 46; ERp46; FLJ21353; FLJ90810; HCC2; HCC-2; MGC3178; PC-TRP; PDIA15; Plasma cell-specific thioredoxin-related protein; protein disulfide isomerase family A, member 15; STRF8; thiore; thioredoxin domain containing 5; thioredoxin domain containing 5 (endoplasmic reticulum); thioredoxin domain-containing protein 5; thioredoxin related protein; thioredoxin-like protein p46; Tlp46; TXNDC5; UNQ364; UNQ364/PRO700 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 81567 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NBS9 | |
| Txndc5 | |
| Recombinant protein corresponding to Human TXNDC5. Recombinant protein control fragment (Product #RP-95032). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction