missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TXNDC5 Monoclonal Antibody (2E7/7), PerCP
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA545307
This item is not returnable.
View return policy
Description
1 μg/mL of MA5-45307 was sufficient for detection of EndoPDI in 20 μg of heat shocked HeLa cell lysate by colorimetric immunoblot analysis using Goat anti -mouse IgG:HRP as the secondary antibody. Detects approximately 48kDa.
This gene encodes a protein-disulfide isomerase. Its expression is induced by hypoxia and its role may be to protect hypoxic cells from apoptosis. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
| TXNDC5 | |
| Monoclonal | |
| 1 mg/mL | |
| 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4 | |
| Q8NBS9 | |
| Txndc5 | |
| Synthesized peptide - 12 amino acids long (ADGEDGQDPHSK) corresponding to residues 52 - 63 of the EndoPDI protein. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 2E7/7 | |
| PerCP | |
| Txndc5 | |
| AL022641; ENDOPDI; endoplasmic reticulum protein ERp46; endoplasmic reticulum resident protein 46; endothelial protein disulphide isomerase; ER protein 46; ERp46; FLJ21353; FLJ90810; HCC2; HCC-2; MGC3178; PC-TRP; PDIA15; Plasma cell-specific thioredoxin-related protein; protein disulfide isomerase family A, member 15; STRF8; thiore; thioredoxin domain containing 5; thioredoxin domain containing 5 (endoplasmic reticulum); thioredoxin domain-containing protein 5; thioredoxin related protein; thioredoxin-like protein p46; Tlp46; TXNDC5; UNQ364; UNQ364/PRO700 | |
| Mouse | |
| Protein G | |
| RUO | |
| 81567 | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction