missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TXNDC5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98558-100ul
This item is not returnable.
View return policy
Description
TXNDC5 Polyclonal antibody specifically detects TXNDC5 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| TXNDC5 | |
| Polyclonal | |
| Western Blot 1:5000-1:20000, Immunocytochemistry/ Immunofluorescence 1:100-1:500, Immunoprecipitation 0.5-1 μL/mg of lysate, Immunohistochemistry-Paraffin 1:1000-1:10000 | |
| ENDOPDI, ERP46, FLJ90810, HCC-2, member 15, MGC3178, PDIA15, thioredoxin domain containing 5, thioredoxin domain containing 5 (endoplasmic reticulum), thioredoxin domain-containing protein 5, thioredoxin related protein, TLP46 | |
| Produced in rabbits immunized with E. coli-derived Human TXNDC5 fragment. | |
| 100 μg | |
| Apoptosis | |
| 81567 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction