missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TXNDC5 Antibody (2E7/7), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22414
This item is not returnable.
View return policy
Description
TXNDC5 Monoclonal specifically detects TXNDC5 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications
| TXNDC5 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:200, ELISA, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500 | |
| ENDOPDI, ERP46, FLJ90810, HCC-2, member 15, MGC3178, PDIA15, thioredoxin domain containing 5, thioredoxin domain containing 5 (endoplasmic reticulum), thioredoxin domain-containing protein 5, thioredoxin related protein, TLP46 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Detects approx 48kDa. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| 2E7/7 | |
| Unconjugated | |
| Q8NBS9 | |
| TXNDC5 | |
| Synthesized peptide - 12 amino acids long (ADGEDGQDPHSK) corresponding to residues 52 - 63 of the EndoPDI protein | |
| 0.1 mg | |
| Apoptosis | |
| 81567 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction