missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TXNDC3 Polyclonal antibody specifically detects TXNDC3 in Human, Mouse samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | TXNDC3 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS, 50% glycerol, pH7.3 |
| Gene Alias | CILD6Sptrx-2, EC 2.1.1.31, EC 2.5.1.6, NM23-H8, NME8thioredoxin domain-containing protein 3, Spermatid-specific thioredoxin-2, sperm-specific thioredoxin 2, SPTRX2sptrx-2, thioredoxin domain containing 3 (spermatozoa) |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 299-588 of human NME8 (NP_057700.3). |
| Purification Method | Affinity purified |
| Quantity | 100 μg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?