missing translation for 'onlineSavingsMsg'
Learn More

TXNDC12 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18323756 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18323756 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18323756 Supplier Novus Biologicals Supplier No. NBP310018100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TXNDC12 Polyclonal specifically detects TXNDC12 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TXNDC12
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS buffer, 2% sucrose
Gene Alias AG1, AGR1, anterior gradient homolog 1, EC 1.8.4.2, endoplasmic reticulum protein ERp19, Endoplasmic reticulum resident protein 18, Endoplasmic reticulum resident protein 19, endoplasmic reticulum thioredoxin superfamily member, 18 kDa, ER protein 18, ER protein 19, ERP16, ERp18, ERP18AG1, ERP19, hAG-1, hTLP19, PDIA16, protein disulfide isomerase family A, member 16, thioredoxin domain containing 12 (endoplasmic reticulum), thioredoxin domain-containing protein 12, Thioredoxin-like protein p19, TLP19, TLP19hTLP19, TXNDC12 thioredoxin domain containing 12 (endoplasmic reticulum)
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the N terminal region of human TXNDC12 (NP_056997). Peptide sequence METRPRLGATCLLGFSFLLLVISSDGHNGLGKGFGDHIHWRTLEDGKKEA
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51060
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.