missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TXNDC12 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93457-0.1ml
This item is not returnable.
View return policy
Description
TXNDC12 Polyclonal antibody specifically detects TXNDC12 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| TXNDC12 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:100, Immunohistochemistry-Paraffin | |
| AG1, AGR1, anterior gradient homolog 1, EC 1.8.4.2, endoplasmic reticulum protein ERp19, Endoplasmic reticulum resident protein 18, Endoplasmic reticulum resident protein 19, endoplasmic reticulum thioredoxin superfamily member, 18 kDa, ER protein 18, ER protein 19, ERP16, ERp18, ERP18AG1, ERP19, hAG-1, hTLP19, PDIA16, protein disulfide isomerase family A, member 16, thioredoxin domain containing 12 (endoplasmic reticulum), thioredoxin domain-containing protein 12, Thioredoxin-like protein p19, TLP19, TLP19hTLP19, TXNDC12 thioredoxin domain containing 12 (endoplasmic reticulum) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 93-172 of human TXNDC12 (NP_056997.1). NLEDEEEPKDEDFSPDGGYIPRILFLDPSGKVHPEIINENGNPSYKYFYVSAEQVVQGMKEAQERLTGDAFRKKHLEDEL | |
| 0.1 mL | |
| Signal Transduction | |
| 51060 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction