missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TXNDC Antibody (OTI3H5), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74725H
This item is not returnable.
View return policy
Description
TXNDC Monoclonal specifically detects TXNDC in Human, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| TXNDC | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| TMX1 | |
| Full length recombinant protein of human TXNDC (NP_110382) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI3H5 | |
| Western Blot, Immunohistochemistry | |
| DKFZp564E1962, PDIA11, protein disulfide isomerase family A, member 11, thioredoxin domain containing 1, thioredoxin domain-containing, Thioredoxin domain-containing protein 1, thioredoxin-related transmembrane protein 1, TMXTXNDC, Transmembrane Trx-related protein, TXNDC1thioredoxin domain containing | |
| Mouse | |
| Affinity Purified | |
| Signal Transduction | |
| 81542.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction