missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TXLNG Polyclonal Antibody

TXLNG Polyclonal Antibody
Brand: Invitrogen PA544156
This item is not returnable.
View return policy
Description
TXLNG Polyclonal Antibody for Western Blot
Specifications
| TXLNG | |
| Polyclonal | |
| Unconjugated | |
| TXLNG | |
| 4932441K18Rik; CXHXORF15; CXorf15; ELRG; environmental lipopolysaccharide-responding gene protein; environmental LPS-responding; factor inhibiting activating transcription factor 4 (ATF4)-mediated transcription; factor inhibiting ATF4-mediated transcription; FIAT; gamma-taxilin; lipopolysaccharide specific response-5 protein; lipopolysaccharide-responsive gene protein; Lipopolysaccharide-specific response protein 5; Lrg; Lrp; LSR5; taxilin gamma; TXLNG; TXLNGX | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 55787 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9NUQ3 | |
| TXLNG | |
| Synthetic peptide directed towards the N-terminal of human TXLNG (aa 51-100). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction