Learn More
Thermo Scientific™ TX-750 Round Bucket Adapters
Product Code. 11814471 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity:
1.5/2 mL Conical Microtube
100 mL Round-bottom Open-top Tube
14 mL Conical Urine Tube
15 mL Conical Tube
25 mL Universal Conical/Skirted Tube
250 mL Bottle
250 mL Corning Conical Bottle
5 mL RIA/Round-bottom Tube (w/o cap)
5/7 mL Blood Collection Tube
5/7 mL Round-bottom Tube (w/o cap) w/Decanting Aid
50 mL Conical Tube
50 mL Round-bottom Tube
84 x 15 mL Blood Collection Tube
Unit Size:
Pack of 4
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity unitSize
11814471 1.5/2 mL Conical Microtube Pack of 4
10491445 50 mL Conical Tube Pack of 4
10287707 15 mL Conical Tube Pack of 4
11894451 250 mL Bottle Pack of 4
10595494 100 mL Round-bottom Open-top Tube Pack of 4
11824461 50 mL Round-bottom Tube Pack of 4
11844461 25 mL Universal Conical/Skirted Tube Pack of 4
11864461 14 mL Conical Urine Tube Pack of 4
11874461 84 x 15 mL Blood Collection Tube Pack of 4
11884461 5/7 mL Blood Collection Tube Pack of 4
11894461 5 mL RIA/Round-bottom Tube (w/o cap) Pack of 4
11804471 5/7 mL Round-bottom Tube (w/o cap) w/Decanting Aid Pack of 4
11953759 250 mL Corning Conical Bottle Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Product Code. 11814471 Supplier Thermo Scientific™ Supplier No. 75003733
In Stock
Add to basket
Add to basket
Add functionality to your rotor with round bucket adapters, which provide support for round-bottom and conical tubes and bottles, microtubes, and blood collection tubes.
Add functionality to your rotor with Thermo Scientific™ TX-750 Round Bucket Adapters, which provide support for round-bottom and conical tubes and bottles, microtubes and blood collection tubes.
Adapters will hold round-bottom and conical tubes and bottles, microtubes and blood collection tubes.
TRUSTED_SUSTAINABILITY
Specifications
Type Round Bucket Adapter
Tube Type Microtube, Conical
Capacity 1.5/2 mL Conical Microtube
For Use With (Equipment) Round-Bottom and Conical Tube and Bottle, Microtube and Blood Collection Tube
Tube Size 1.5/2 mL