Learn More
Thermo Scientific™ TX-750 Rectangular Bucket Adapters
Product Code. p-4527794 Shop All Thermo Scientific Products
Click to view available options
Capacity:
15 mL Conical Tubes
50 mL Conical Tubes
Unit Size:
Pack of 4
Product Code. 11854481
Brand: Thermo Scientific™ 75003684
Use Thermo Scientific™ TX-750 Rectangular Bucket Adapters with TX-750 Rectangular Rotor Buckets.
Use Thermo Scientific™ TX-750 Rectangular Bucket Adapters with TX-750 Rectangular Rotor Buckets.
Ordering Information:
Order rectangular buckets (75003614), Thermo Scientific™ ClickSeal™ Biocontainment Lids (75003615) and TX-750 Rotor (75003607) separately. Contact your sales representative for more information.
TRUSTED_SUSTAINABILITY
Specifications
Type Tube Adapter
Capacity 15 mL Conical Tubes