missing translation for 'onlineSavingsMsg'
Learn More

TWS119, MedChemExpress

Product Code. 16450417
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16450417

Brand: Medchem Express HY10590

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress TWS119 is a specific inhibitor of GSK-3β, with an IC50 of 30 nM, and activates the wnt/β-catenin pathway.

TWS119 is a specific inhibitor of GSK-3β, with an IC50 of 30 nM, and activates the wnt/β-catenin pathway.

Specifications

Chemical Name or Material TWS119
CAS 601514-19-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C18H14N4O2
Quantity 50 mg
Solubility Information DMSO : ≥ 50 mg/mL (157.07 mM) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES OC1=CC=CC(OC2=C3C(NC(C4=CC=CC(N)=C4)=C3)=NC=N2)=C1
Molecular Weight (g/mol) 318.33
Formula Weight 318.33
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.