missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ Two-Tier Polypropylene Test Tube Racks
Shop All SP Scienceware Products
Click to view available options
Holds:
Tubes (13 to 16 mm)
Tubes (16 to 20 mm)
Diameter (Metric) Opening:
16 mm
20 mm
Unit Size:
Each
Description
Designed to withstand elevated water temperatures for incubation.
- Hold serum vials, RIA vials, T3 or T4 reaction vials, or any other vial or tube of same dimensions
- Won't rust or corrode
- Lightweight
- Autoclavable
- Polypropylene construction
- Includes endplate handles
Specifications
Specifications
| Includes | Integral endplate handles |
| Product Type | Tube Rack (2-tier) |
| Array | 4 x 10 |
| Autoclavable | Autoclavable |
| Holds | Tubes (13 to 16 mm) |
| Material | Polypropylene |
| Positions | 40 |
| Shape | Rectangular |
| Quantity | 1 Each |
| Dimensions (L x W x H) | 254 x 111 x 89mm |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction