missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Standard Two-Prong Extension Clamps

Product Code. 11735419 Shop All Cole-Parmer Products
Change view
Click to view available options
Diameter (Metric):
75 mm
92 mm
95 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
11735419 92 mm Each
11745419 95 mm Each
11939718 75 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11735419 Supplier Cole-Parmer™ Supplier No. 0802144
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Securely hold burets, thermometers, beakers, and flask necks with these extension clamps. Standard Two-Prong Extension Clamps are deal for holding straight-sided apparatus. Dual adjustable clamps are offered in either stainless steel or nickel plated zinc.

Choose single adjustment style for standard use where one prong or finger is adjusted using a wing nut, while the other prong remains stationary or for a more versatile clamp choose the dual adjustment style.

  • Both prongs are easily adjusted over a wider range
  • Available in medium and large sizes
  • Supplied with non-slip vinyl sleeves or for temperatures above 100°C, fiberglass covers
  • An additional holder must be purchased in order to attach clamps to lab-frames, support stands or other apparatus

Specifications

Quantity 1 Ea.
Type Utility Clamp
Material Nickel-plated Zinc
For Use With (Application) Ideal for holding straight-sided apparatus such as burets, thermometers, beakers, and flask necks
Diameter (Metric) 92 mm
Length (Metric) 233 mm
Size Large
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.