missing translation for 'onlineSavingsMsg'
Learn More

Ohaus™ Two Block Digital Dry Block Heater HB2DG

Product Code. 15712709 Shop All Ohaus Products
Change view
Click to view available options
Controller Type:
Digital
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Controller Type unitSize
15712709 Digital Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15712709 Supplier Ohaus™ Supplier No. 30392082
  • ONLINE EXCLUSIVE PRICE
    £1041.21
    £914.96 / Pack of 1
    Save £126.25
    12% Off
View all promotions
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

OHAUS™ Two Block Digital Dry Block Heater processes more samples while delivering precise and reliable results.

  • Designed for applications that require repeatable results and superior temperature stability
  • Multi-purpose units are ideal for incubation and activation of cultures, enzyme reactions, immunoassays, melting/boiling points, and a wide variety of other laboratory procedures
  • Integral support rod holder with locking knob accepts optional external temperature probe kit
  • External temperature probe kit monitors actual block or sample temperature
  • Each of the five available models accepts separate interchangeable modular blocks (available separately) accommodating various tube sizes from 0.2 mL microtubes to 50 mL centrifuge tubes
  • Each block has a thermometer well for measuring block temperature
  • Efficient heating due to close tube-and-block contact for maximum heat retention
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance TUV
Controller Type Digital
Description Dry Block Heater, 2 Block, HB2DG, EU
Temperature Range (Metric) 5°C to 120°C
Type Dry Bath Block
Heat Up Time 50 min
Model HB2DG
No. of Blocks 2
Temperature Stability ±0.1°C
Temperature Uniformity ±0.1°C at 37°C
Uniformity ± 0.2°C at 37°C
Dimensions (L x W) 39.10 cm x 20.30 cm
Dimensions (L x W x H) 39.1 x 20.3 x 8.9 cm
Length (English) 15.40 in.
Length (Metric) 39.10 cm
Width (English) 8.00 in.
Width (Metric) 20.30 cm
Height (English) 3.50 in.
Height (Metric) 8.900 cm
Depth (English) 15.40 in.
Depth (Metric) 39.10 cm
Weight (Metric) 2.8 kg
Electrical Requirements 230V, 0.92A, 50/60Hz
Amperage 0.92A
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.