missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TWIST2 Mouse anti-Human, Mouse, Rat, Clone: 1B12D11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66544-1-IG-20UL
This item is not returnable.
View return policy
Description
Binds to the E-box consensus sequence 5′-CANNTG-3′ as a heterodimer and inhibits transcriptional activation by MYOD1, MYOG, MEF2A and MEF2C. Also represses expression of proinflammatory cytokines such as TNFA and IL1B. Involved in postnatal glycogen storage and energy metabolism. Inhibits the premature or ectopic differentiation of preosteoblast cells during osteogenesis, possibly by changing the internal signal transduction response of osteoblasts to external growth factors.Specifications
| TWIST2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P97831, Q8WVJ9, Q9D030 | |
| Twist2 | |
| TWIST2 Fusion Protein Ag2348 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG1 |
| Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1B12D11 | |
| Unconjugated | |
| TWIST2 | |
| bHLHa39, Dermis expressed protein 1, Dermo 1, DERMO1, twist homolog 2 (Drosophila), Twist related protein 2, TWIST2 | |
| Mouse | |
| Protein G | |
| RUO | |
| 117581, 13345, 59327 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction