missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TWIST2 Mouse anti-Human, Clone:1A11D9, Abnova™
Mouse monoclonal antibody raised against recombinant human TWIST2.
Brand: Abnova MAB17845.100ug
This item is not returnable.
View return policy
Description
Basic helix-loop-helix (bHLH) transcription factors have been implicated in cell lineage determination and differentiation. The protein encoded by this gene is a bHLH transcription factor and shares similarity with another bHLH transcription factor, Twist. It is thought that during osteoblast development this protein may inhibit osteoblast maturation and maintain cells in a preosteoblast phenotype. [provided by RefSeq]Specifications
| TWIST2 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:100-1:500) The optimal working dilution should be determined by the end user. |
|
| TWIST2 | |
| Mouse | |
| 100 μg | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Western Blot | |
| 1A11D9 | |
| twist homolog 2 (Drosophila) | |
| In PBS (0.05% sodium azide) | |
| DERMO1, MGC117334, bHLHa39 | |
| Recombinant protein corresponding to amino acids 1-160 of human TWIST2 from E. coli. | |
| 117581 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction