missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Twist-1 Antibody (927403), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB6230CL1
This item is not returnable.
View return policy
Description
Twist-1 Monoclonal antibody specifically detects Twist-1 in Human samples. It is validated for Western Blot,Flow Cytometry (Intracellular Staining),CyTOF
Specifications
| Twist-1 | |
| Monoclonal | |
| CoraFluor 1 | |
| acrocephalosyndactyly 3, ACS3, B-HLH DNA binding protein, BHLHA38, bHLHa38BPES3, Class A basic helix-loop-helix protein 38, CRS1, H-twistblepharophimosis, epicanthus inversus and ptosis 3, SCSBPES2, TWIST, twist homolog 1 (Drosophila), TWIST homolog of drosophila, twist-related protein 1 | |
| E.coli-derived recombinant human Twist-1, Met1-His202, Accession # Q15672 | |
| 0.1 mL | |
| Cancer | |
| 7291 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Western Blot, Flow Cytometry (Intracellular Staining), CyTOF | |
| 927403 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction