missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Twinkle Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18793-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a hexameric DNA helicase which unwinds short stretches of double-stranded DNA in the 5′ to 3′ direction and, along with mitochondrial single-stranded DNA binding protein and mtDNA polymerase gamma, is thought to play a key role in mtDNA replication. The protein localizes to the mitochondrial matrix and mitochondrial nucleoids. Mutations in this gene cause infantile onset spinocerebellar ataxia (IOSCA) and progressive external ophthalmoplegia (PEO) and are also associated with several mitochondrial depletion syndromes. Alternative splicing results in multiple transcript variants encoding distinct isoforms.Specifications
| Twinkle | |
| Polyclonal | |
| Unconjugated | |
| TWNK | |
| ATXN8, C10orf2, IOSCA, PEO, PEO1, PEOA3, SANDO, SCA8, Twinkle, Twinkle protein, mitochondrial, TWINL | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 56652 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q96RR1 | |
| TWNK | |
| Twinkle Fusion Protein Ag6628 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction