missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TWF2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562858
This item is not returnable.
View return policy
Description
Immunogen sequence: HQTGIHATEE LKEFFAKARA GSVRLIKVVI EDEQLVLGAS QEPVGRWDQD YDRAVLPLLD AQQPCYLLYR Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.
TWF2 encodes a protein that was identified by its interaction with the catalytic domain of protein kinase C-zeta. The encoded protein contains an actin-binding site and an ATP-binding site. It is most closely related to twinfilin, a conserved actin monomer-binding protein.
Specifications
| TWF2 | |
| Polyclonal | |
| Unconjugated | |
| TWF2 | |
| A6 related protein; A6r; A6-related; A6-related protein; A6RP; AU014993; hA6RP; mA6RP; MSTP011; protein tyrosine kinase 9 related protein; Protein tyrosine kinase 9-like; protein tyrosine kinase 9-like (A6-related protein); PTK 9L; PTK9 protein typrotein tyrosine kinase 9-like (A6-related protein); Ptk9l; PTK9L protein tyrosine kinase 9-like (A6-related protein); Ptk9r; TWF 2; Twf2; Twinfilin 2; twinfilin actin binding protein 2; twinfilin actin-binding protein 2; twinfilin, actin-binding protein, homolog 2; twinfilin, actin-binding protein, homolog 2 (Drosophila); twinfilin-1-like protein; Twinfilin2; Twinfilin-2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11344 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6IBS0 | |
| TWF2 | |
| Recombinant protein corresponding to Human TWF2. Recombinant protein control fragment (Product #RP-101485). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction