Learn More
TWF2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA589341
Description
Immunogen sequence: MAHQTGIHAT EELKEFFAKA RAGSVRLIKV VIEDEQLVLG ASQEPVGRWD QDYDRAVLPL LDAQQPCYLL YRLDSQNAQG FEWLFLAWSP DNSPVRLKML YAATRATVKK EFGGGHIKDE LFGTVKDDLS FAGYQKHLSS CAAPAPLTSA ERELQQIRIN EVKTEISVES KHQTLQGLAF PLQPEAQRAL QQLKQKMVNY IQMKLDLERE TIELVHTEPT DVAQLPSRVP RDAARYHFFL YKHTHEGDPL ESVVFIYSMP GYKCSIKERM LYSSCKSRLL DSVEQDFHLE IAKKIEIGDG AELTAEFLYD EVHPKQHAFK QAFAKPKGPG GKRGHKRLIR GPGENGDDS; Positive Samples: Jurkat, 293T, A-431, Mouse heart, Mouse thymus; Cellular Location: Cell projection, Cytoplasm, cytoskeleton, perinuclear region, stereocilium
TWF2 encodes a protein that was identified by its interaction with the catalytic domain of protein kinase C-zeta. The encoded protein contains an actin-binding site and an ATP-binding site. It is most closely related to twinfilin, a conserved actin monomer-binding protein.
Specifications
| TWF2 | |
| Polyclonal | |
| Unconjugated | |
| TWF2 | |
| A6 related protein, A6r, A6-related, A6-related protein, A6RP, AU014993, hA6RP, mA6RP, MSTP011, protein tyrosine kinase 9 related protein, Protein tyrosine kinase 9-like, protein tyrosine kinase 9-like (A6-related protein), PTK 9L, PTK9 protein typrotein tyrosine kinase 9-like (A6-related protein), Ptk9l, PTK9L protein tyrosine kinase 9-like (A6-related protein), Ptk9r, TWF 2, Twf2, Twinfilin 2, twinfilin actin binding protein 2, twinfilin actin-binding protein 2, twinfilin, actin-binding protein, homolog 2, twinfilin, actin-binding protein, homolog 2 (Drosophila), twinfilin-1-like protein, Twinfilin2, Twinfilin-2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 11344, 23999, 684352 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunoprecipitation, Western Blot, Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300, pH 7.3 | |
| Q6IBS0, Q9Z0P5 | |
| TWF2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-349 of human TWF2 (NP_009215.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.