missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polysorbate 80, Thermo Scientific Chemicals
52.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
54.04 GBP valid until 2024-04-30
Use promo code "21741" to get your promotional price.
Use promo code "22914" to get your promotional price.
54.04 GBP valid until 2024-04-30
Use promo code "21741" to get your promotional price.
Click to view available options
Quantity:
250 mL
1 L
2.5 L
10 L
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Yellow to Amber |
Content And Storage | Indoors +20°C |
Density | 1.0800 g/mL |
Molecular Formula | (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6 |
MDL Number | MFCD00082107 |
Quantity | 1 L |
Packaging Type | Glass bottle |
InChI Key | HDTIFOGXOGLRCB-KTKRTIGZNA-N |
SMILES | CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO |
IUPAC Name | 2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'