missing translation for 'onlineSavingsMsg'
Learn More

Polysorbate 80

Product Code. 10048651 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
10 L
Unit Size:
1 L
10 L
2.5 L
250mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10048651 2.5 L 2.5 L
10389150 250 mL 250mL
10410911 1 L 1 L
10246943 10 L 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10048651 Supplier Thermo Scientific Acros Supplier No. 278630025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Polysorbate 80, CAS Number-9005-65-6, Polyoxyethylene(20)sorbitan monooleate, 1L, 2mg KOH/g max., Yellow to Amber, 1.0800g/mL, 5ppm max., >110 deg.C, 65 to 80mg KOH/g, Authentic, 15, 7703, MFCD00082107, Glass bottle, Viscous Liquid At 25 deg.C, Indoors +20 deg.C, 1.4710 to 1.4730, 45 to 55mg KOH/g | CAS: 9005-65-6 | (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6 | 604.82 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9005-65-6
Synonym Polyoxyethylene(20)sorbitan monooleate

Specifications

Color Yellow to Amber
Content And Storage Indoors +20°C
Density 1.0800 g/mL
Quantity 2.5 L
Molecular Formula (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6
MDL Number MFCD00082107
Packaging Type Glass bottle
InChI Key HDTIFOGXOGLRCB-KTKRTIGZNA-N
SMILES CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
IUPAC Name 2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate
Molecular Weight (g/mol) 604.82
Solubility Solubility in water: 100g/L. Other solubilities: 100g/L isopropyl alcohol, 10g/L xylene, insoluble in propylene glycol and mineral oil
Form Viscous Liquid
Chemical Name or Material Polysorbate 80
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.