missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polysorbate 20, Thermo Scientific Chemicals
£16.40 - £347.00
Chemical Identifiers
CAS | 9005-64-5 |
---|---|
Molecular Formula | C26H50O10 |
Synonym | Polyoxyethylene(20)sorbitan monolaurate |
Product Code | Brand | Quantity | Packaging Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging Type | Price | Quantity & Availability | ||||
10700941
|
Thermo Scientific Acros
233360050 |
5 mL | Glass Bottle |
£16.40
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10246910
|
Thermo Scientific Acros
233362500 |
250 mL | Glass Bottle |
£29.00
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10491081
|
Thermo Scientific Acros
233360010 |
1 L | Glass Bottle |
£61.30
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10503874
|
Thermo Scientific Acros
233360051 |
5 L | Plastic Bottle |
£160.00
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10698313
|
Thermo Scientific Acros
233360250 |
25 L | Plastic drum |
£347.00
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
9005-64-5 | |
C26H50O10 | |
Polyoxyethylene(20)sorbitan monolaurate |
Specifications
9005-64-5 | |
1.1000 g/mL | |
Solubility in water: 100g/L. Other solubilities: 100g/L isopropylalcohol, xylene, insoluble in mineral oil, 10g/L propylene glycol | |
Liquid |
Colorless to Yellow | |
C26H50O10 | |
Polyoxyethylene(20)sorbitan monolaurate | |
Polysorbate 20 |
RUO – Research Use Only