missing translation for 'onlineSavingsMsg'
Learn More

TWEEN 20 (50% Solution)

TWEEN 20 (50% Solution)

Brand:  Invitrogen 003005

Quantity 20mL

Product Code. 10642074

  • £85.75 / Pack of 1
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

May be added to TBS or PBS at low concentrations, to reduce non-specific binding in ELISAs and IHC.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

20mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.