missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TWEAK/TNFSF12 Antibody (MM0590-10T24), mFluor Violet 610 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
TWEAK/TNFSF12 Monoclonal antibody specifically detects TWEAK/TNFSF12 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | TWEAK/TNFSF12 |
| Classification | Monoclonal |
| Host Species | Mouse |
| Immunogen | Human recombinant TWEAK/TNFSF12 |
| Purification Method | Protein G purified |
| Quantity | 0.1 mL |
| Research Discipline | Angiogenesis, Apoptosis, Cancer, Tumor Suppressors |
| Primary or Secondary | Primary |
| Isotype | IgG1 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?