missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TWEAK/TNFSF12 Antibody (C06-3C8), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11625-0.1mg
This item is not returnable.
View return policy
Description
TWEAK/TNFSF12 Monoclonal antibody specifically detects TWEAK/TNFSF12 in Human samples. It is validated for ELISA
Specifications
| TWEAK/TNFSF12 | |
| Monoclonal | |
| Unconjugated | |
| APO3 ligand, APO3LMGC20669, DR3LGTWEAKAPO3/DR3 ligand, MGC129581, TNF-related weak inducer of apoptosis, tumor necrosis factor (ligand) superfamily, member 12, tumor necrosis factor ligand superfamily member 12 | |
| E. coli derived recombinant protein corresponding to aa 97-249 of human TWEAK | |
| 0.1 mg | |
| Angiogenesis, Apoptosis, Cancer, Tumor Suppressors | |
| 8742 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
| ELISA | |
| C06-3C8 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction