missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TWEAK R/TNFRSF12 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-34499AF594
This item is not returnable.
View return policy
Description
TWEAK R/TNFRSF12 Polyclonal antibody specifically detects TWEAK R/TNFRSF12 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| TWEAK R/TNFRSF12 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 594 | |
| CD266, CD266 antigen, FGF-inducible 14, Fibroblast growth factor-inducible immediate-early response protein 14, FN14tumor necrosis factor receptor superfamily member 12A, tumor necrosis factor receptor superfamily, member 12A, TweakR, Tweak-receptor, type I transmembrane protein Fn14 | |
| Antibody was raised against a human IgG1 Fc-tagged Fn14 protein. | |
| 0.1 mL | |
| Angiogenesis, Apoptosis, Biologically Active Proteins, Cancer, Tumor Suppressors | |
| 51330 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction