missing translation for 'onlineSavingsMsg'
Learn More

TWEAK R/TNFRSF12 Antibody (314615) [PerCP], Novus Biologicals™

Product Code. 30735965 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30735965 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30735965 Supplier Novus Biologicals Supplier No. FAB1610C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

TWEAK R/TNFRSF12 Monoclonal antibody specifically detects TWEAK R/TNFRSF12 in Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen TWEAK R/TNFRSF12
Applications Western Blot
Classification Monoclonal
Clone 314615
Conjugate PerCP
Formulation PBS
Gene Alias CD266, CD266 antigen, FGF-inducible 14, Fibroblast growth factor-inducible immediate-early response protein 14, FN14tumor necrosis factor receptor superfamily member 12A, tumor necrosis factor receptor superfamily, member 12A, TweakR, Tweak-receptor, type I transmembrane protein Fn14
Host Species Rat
Immunogen E. coli-derived recombinant mouse TWEAK R/TNFRSF12, Glu28-Trp79, Accession # Q9CR75
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Apoptosis, Biologically Active Proteins, Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 51330
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.