missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TWEAK R (CD266/TNFRSF12A) Mouse ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ EMTNFRSF12AX10
Additional Details : Weight : 2.50000kg
Description
TWEAK is a member of the TNF family and was first described as a weak inducer of apoptosis in demonstrate that this relatively small molecule is a fully transformed cell lines. Recently, a receptor for TWEAK was isolated by expression cloning from a HUVEC cell cDNA library. TweakR is a small protein belonging to TNF receptor super family. TweakR is capable of intiating a proliferative signal by binding to TRAF1, 2, and 3. TweakR mRNA is upregulated during tissue regeneration in the aorta and liver. It may also play a role in vascular cell migration and angiogenesis.Specifications
Q9CR75 | |
95 pg/mL | |
ELISA | |
Colorimetric Microplate Reader | |
TNFRSF12A | |
<10% | |
HRP | |
RUO | |
2°C to 8°C | |
1 hr 20 min |
102.2 to 25,000 pg/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
27279 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
10 x 96 Tests | |
Plasma, 33 μL|Serum, 33 μL|Supernatant, 100 μL | |
CD266 | |
4 hr 45 min |