Learn More
Brannan™ Twaddell Hydrometers
GREENER_CHOICE
Product Code. 12859177
Click to view available options
:
Specific Gravity Range: 74 to 102 Tw
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 12859177
Brand: Brannan™ 10/104/0
Only null left
Add to basket
Edge
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
For reporting the measured specific gravity of a liquid relative to water. Brannan™ Twaddell Hydrometers are manufactured from soda glass, steel shot ballast and synthetic wax resin.
For reporting the measured specific gravity of a liquid relative to water. Brannan™ Twaddell Hydrometers are manufactured from soda glass, steel shot ballast and synthetic wax resin.
  • 160 mm in length
  • With single degree divisions
  • Adjusted for use at 15.6°C (60°F)
  • For medium surface tension liquids
  • Available for numerous Twaddell ranges