missing translation for 'onlineSavingsMsg'
Learn More

Brannan™ Twaddell Hydrometers

Product Code. 12829177
Change view
Click to view available options
Specific Gravity Range:
0 to 24 Tw
24 to 48 Tw
74 to 102 Tw
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Specific Gravity Range unitSize
12829177 0 to 24 Tw Each
12839177 24 to 48 Tw Each
31488168 74 to 102 Tw -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12829177 Supplier Brannan™ Supplier No. 10/101/0
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

For reporting the measured specific gravity of a liquid relative to water. Brannan™ Twaddell Hydrometers are manufactured from soda glass, steel shot ballast and synthetic wax resin.

For reporting the measured specific gravity of a liquid relative to water. Brannan™ Twaddell Hydrometers are manufactured from soda glass, steel shot ballast and synthetic wax resin.

  • 160 mm in length
  • With single degree divisions
  • Adjusted for use at 15.6°C (60°F)
  • For medium surface tension liquids
  • Available for numerous Twaddell ranges

Specifications

Quantity 1 Ea.
Type Twaddle Hydrometer
Accuracy ±1 Tw
Increments 1 Tw
Length (Metric) 160 mm
Ballast Type Steel Shot
Baume Range 0 to 24 Tw
For Use With (Application) For Reporting The Measured Specific Gravity Of A Liquid Relative To Water
Specific Gravity Range 0 to 24 Tw
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.