missing translation for 'onlineSavingsMsg'
Learn More

TW-37, MedChemExpress

Product Code. 16424928
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16424928 10 mg 10mg
16411177 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16424928 Supplier Medchem Express Supplier No. HY12020
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress TW-37 is a potent Bcl-2 inhibitor with Ki values of 260, 290 and 1110 nM for Mcl-1, Bcl-2 and Bcl-xL, respectively.

TW-37 is a potent Bcl-2 inhibitor with Ki values of 260, 290 and 1110 nM for Mcl-1, Bcl-2 and Bcl-xL, respectively.

Specifications

Chemical Name or Material TW-37
CAS 877877-35-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C33H35NO6S
Quantity 10 mg
Solubility Information DMSO : ≥ 42 mg/mL (73.21 mM)
SMILES O=C(C1=CC(CC2=CC=CC=C2C(C)C)=C(C(O)=C1O)O)NC3=CC=C(C=C3)S(=O)(C4=CC=CC=C4C(C)(C)C)=O
Molecular Weight (g/mol) 573.7
Formula Weight 573.7
Percent Purity 96.73%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.