missing translation for 'onlineSavingsMsg'
Learn More

Scie Plas TV100YK-EBSYS TV Modular Vertical Electroblotting System

Product Code. 15865551
Click to view available options
Amperage:
10 to 15 mA
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15865551

Brand: Scie Plas TV100YKEBSYS

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Provides a complete solution within one gel tank for PAGE and electroblotting.

  • Comprises of one standard GRM and one EBGRM, which are interchangeable within the same gel tank, allowing to perform electroblotting independently or as part of a fully integrated system
  • Twin-cassette EBGRM is available in 2 sizes to allow simultaneous transfer of 2 polyacrylamide gels
  • Plate-electrodes are only 6cm apart which maximise the intensity of the electric field, allowing rapid transfer in as little as 1 hour
  • Colour-coded electroblotting cassettes corresponding to the anode and cathode ensures that transfer is performed in the correct orientation
  • Fibre pads compress the gel against the membrane for uniform transfer
  • Twin-plate standard GRM designed specifically for SDS-PAGE techniques requiring self-cast gels
  • Casting base and cooling are included as standard act in tandem with glass plate stops within the GRM to lock the plates firmly in position on the casting base

Specifications

Type TV Modular Vertical Electroblotting System
Includes Twin-plate mini-gel unit with GRM and cooled gel tank, with built-in cooling coil and quick-fit tubing, lid, 2 x (10W x 10cmH) plain glass plates, 2 x (10 x 10cm) notched glass plates, 1 x dummy plate, 4 x 1mm thick spacers, 2 x spacer aligners and 2 x 1mm thick 12-sample combs, casting base and 2 x silicone seals; plus EBGRM, 2 x electroblotting cassettes and 4 x fibre pads
Volume (Metric) Buffer Gel Tank: 1600 mL
Volume (Metric) Chamber Inner Buffer Chamber: 90 mL
Dimensions (L x W x H) W x H: 28 x 18 cm
Voltage 100 to 150 V
Amperage 10 to 15 mA
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.