missing translation for 'onlineSavingsMsg'
Learn More

Scie Plas TV100K Cooled Twin-Plate Mini-Gel Electrophoresis Unit

Ideal solution for standard PAGE applications using either self-cast or precast gels.

Brand:  Scie Plas TV100K


Additional Details : Weight : 0.40000kg

Product Code. 15835551

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • Smaller footprint area
  • Contains cooling coil in the bottom of the gel tank for faster higher voltage electrophoretic separations
  • Enhanced cooling with snap-lock connectors connected to the external chiller unit
  • Compatible with commercially available 10 x 10cm precast gels
  • Gel casting can be done with tape or optional casting base
  • Float glass plates with ground edges provides uniform gel thickness, sample migration and inhibits sample leakage during casting
  • Single screws at either side each gel clamping plate disperse pressure evenly along the height and breadth of the gel as the screws are tightened
  • Low buffer volumes
  • Color coded combs and spacers
  • Recommended power supply is Consort EV243
Specifications

Specifications

Snap-lock connectors for cooling coil
TV100K Cooled Twin-Plate Mini-Gel Electrophoresis Unit
0.1 cm
W x H: 28 x 18 cm
100 to 150 V
1 to 1.75 hr.
W x H: 8 x 8.5 cm
Inner chamber: 90 mL; Gel tank: 1600 mL
15 cm
10 to 15 mA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers