missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TUSC5 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5116271

Product Code. 17137508

  • £463.00 / 400µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TUSC5 Polyclonal Antibody for Western Blot

TUSC5 (tumor suppressor candidate 5), also known as protein located at seventeen-p-thirteen point three 1, LOST1 or IFITMD3 (interferon-induced transmembrane domain-containing protein D3), is a 177 amino acid multi-pass membrane protein that belongs to the CD225 family. Thought to play a role in fat metabolism, TUSC5 is highly expressed in mammary gland, heart, smooth muscle, skeletal muscle and stomach, with lower levels found in lung and brain.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TUSC5
Polyclonal
Unconjugated
TUSC5
BEC-1; brain endothelial cell-derived protein 1; C130069F04Rik; Dispanin subfamily B member 1; DSPB1; IFITMD3; interferon induced transmembrane protein domain containing 3; interferon-induced transmembrane domain-containing protein D3; located at seventeen p thirteen point three 1; Lost1; Protein located at seventeen-p-thirteen point three 1; Trafficking regulator of GLUT4 1; Trarg1; Tumor suppressor candidate 5; tumor suppressor candidate 5 homolog; Tusc5
Rabbit
Antigen Affinity Chromatography, Protein A
RUO
286753
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.4
Q8IXB3
TUSC5
KLH conjugated synthetic peptide between 78-104 amino acids from the Central region of human TUSC5.
400 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.