missing translation for 'onlineSavingsMsg'
Learn More

TUSC4 Antibody, Novus Biologicals™

Product Code. 18337508 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18337508 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18337508 Supplier Novus Biologicals Supplier No. H00010641B02P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

TUSC4 Polyclonal antibody specifically detects TUSC4 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen TUSC4
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation PBS (pH 7.4)
Gene Accession No. AAH21984.1
Gene Alias G21 protein, Gene 21 protein, homologous to yeast nitrogen permease (candidate tumor suppressor), nitrogen permease regulator 2-like protein, nitrogen permease regulator-like 2 (S. cerevisiae), NPR2, NPR2L2810446G01Rik, NPRL2, Tumor suppressor candidate 4NPR2-like protein, TUSC4
Host Species Mouse
Immunogen TUSC4 (AAH21984.1, 1 a.a. - 380 a.a.) full-length human protein. MGSGCRIECIFFSEFHLTLGPKITYQVPEDFISRELFDTVQVYIITKPELQNKLITVTAMEKKLIGCPVCIEHKKYSRNALLFNLGFVCDAQAKTCALEPIVKKLAGYLTTLELESSFVSMEESKQKLVPIMTILLEELNASGRCTLPIDESNTIHLKVIEQRPDPPVAQEYDVPVFTKDKEDFFNSQWDLTTQQILPYIDGFRHIQKISAEADVELNLVRIAIQNLLYYGVVTLVSILQYSNVYCPTPKVQDLVDDKSLQEACLSYVTKQGHKRASLRDVFQLYCSLSPGTTVRDLIGRHPQQLQHVDERKLIQFGLMKNLIRRLQKYPVRVTREEQSHPARLYTGCHSYDEICCKTGMSYHELDERLENDPNIIICWK
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 10641
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.