missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TUSC4 Polyclonal antibody specifically detects TUSC4 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
Specifications
| Antigen | TUSC4 |
| Applications | ELISA, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 |
| Formulation | PBS (pH 7.3), 50% glycerol |
| Gene Alias | G21 protein, Gene 21 protein, homologous to yeast nitrogen permease (candidate tumor suppressor), nitrogen permease regulator 2-like protein, nitrogen permease regulator-like 2 (S. cerevisiae), NPR2, NPR2L2810446G01Rik, NPRL2, Tumor suppressor candidate 4NPR2-like protein, TUSC4 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human TUSC4 (NP_006536.3).,, Sequence:, MGSGCRIECIFFSEFHPTLGPKITYQVPEDFISRELFDTVQVYIITKPELQNKLITVTAMEKKLIGCPVCIEHKKYSRNALLFNLGFVCDAQAKTCALEPIVKKLAGYLTTLELESSFVSMEESKQKLVPIMTILLEELNASGRCTLPIDESNTIHLKVIEQRPDPPVAQEYDVPVFTKDKEDFFNSQWDLTTQQILPYIDGFRHIQKISAEADVELNLV |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?