missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TUSC3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104363
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total TUSC3.
Made up of nearly 146 million bases, chromosome 8 encodes about 800 genes. Translocation of portions of chromosome 8 with amplifications of the c-Myc gene are found in some leukemias and lymphomas, and typically associated with a poor prognosis. Portions of chromosome 8 have been linked to schizophrenia and bipolar disorder. Trisomy 8, also known as Warkany syndrome 2, most often results in early miscarriage but is occasionally seen in a mosaic form in surviving patients who suffer to a varying degree from a number of symptoms including retarded mental and motor development, and certain facial and developmental defects. WRN is a DNA helicase encoded by chromosome 8 and shown defective in those with the early aging disorder Werner syndrome. Chromosome 8 is also associated with Pfeiffer syndrome, congenital hypothyroidism and Waardenburg syndrome.
Specifications
| TUSC3 | |
| Polyclonal | |
| Unconjugated | |
| TUSC3 | |
| AU022242; BC003311; D8S1992; Dolichyl-diphosphooligosaccharide--protein glycosyltransferase subunit TUSC3; M33; Magnesium uptake/transporter TUSC3; MGC13453; MRT22; MRT7; N33; Oligosaccharyl transferase subunit TUSC3; oligosaccharyltransferase 3 homolog A; OST3A; Protein N33; Putative prostate cancer tumor suppressor; tumor suppressor candidate 3; Tusc3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 290783, 7991, 80286 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q13454, Q8BTV1 | |
| TUSC3 | |
| A synthesized peptide derived from human TUSC3(Accession Q13454), corresponding to amino acid residues Y133-A183. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction