missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Turpentine Substitute, Extra Pure, SLR, Fisher Chemical™
Click to view available options
Quantity:
1 L
2.5 L
25 L
Packaging:
Amber glass bottle
HDPE plastic bottle
Metal can
Plastic Coated Glass bottle
Specifications
Specifications
Color | Colorless |
Boiling Point | 149°C |
Packaging | Metal can |
Quantity | 25 L |
Physical Form | Liquid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'