Learn More
Berthold Technologies Turn Table For DeWit Tubes
Product Code. 31777388 Shop All Berthold Technologies Products
Change view
Click to view available options
For Use With (Equipment):
Imaging System
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
31777388 Imaging System Each
1 options
Product Code. 31777388 Supplier Berthold Technologies Supplier No. 5662503
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For NightSHADE imaging systems to hold 18 DeWit tubes for sequential imaging.
  • The turntable for NightSHADE enables sequential and scheduled acquisition of up to 18 DeWitt tubes (in groups of three)
  • Includes complete turntable assembly: base and upper part
Specifications
Type Specimen Tray
For Use With (Application) Imaging of Seedlings
For Use With (Equipment) Imaging System
Voltage 24 V