Learn More
Berthold Technologies Turn Table (Base Only)
Product Code. 31777320 Shop All Berthold Technologies Products
Change view
Click to view available options
For Use With (Equipment):
Imaging System
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
31777320 Imaging System Each
1 options
Product Code. 31777320 Supplier Berthold Technologies Supplier No. 5662504
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Used to image several samples sequentially in NightSHADE imaging systems.
  • Includes only the base of the turntable
  • Complete turntable versions are available for square Petri dishes and DeWitt tubes
Specifications
Type Specimen Tray
For Use With (Application) Imaging of Seedlings
For Use With (Equipment) Imaging System
Voltage 24 V