missing translation for 'onlineSavingsMsg'
Learn More

Hach Lange™ Turbidity Standard 0.50 NTU

Brand:  Hach Lange™ 2698042


Product Code. 10305237

  • £105.67 / 100mL
 Restricted Item
Estimated Shipment: 04-01-2027
Add to basket
This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Turbidity Standard
100 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • StablCal Standard, 0.50 NTU

Signal Word
  • Danger

Hazard Category
  • Skin sensitiser Category 1
  • Respiratory sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • EUH208-Contains Formaldehyde. May produce an allergic reaction.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P285-In case of inadequate ventilation wear respiratory protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P341-IF INHALED: If breathing is difficult, remove victim to fresh air and keep at rest in a position comfortable for breathing.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P363-Wash contaminated clothing before reuse.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.