missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tunicamycin, TRC
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
Specifications
Specifications
| Chemical Name or Material | Tunicamycin |
| CAS | 11089-65-9 |
| Molecular Formula | (CH2)nC29H44N4O16 |
| Quantity | 1 mg |
| SMILES | O=C(NC1=CC=C(Cl)C=C1/C(C2=CC=CC=C2Cl)=N/C(CO)C)CN3CCOCC3 |
| Molecular Weight (g/mol) | 844.95 |
| Formula Weight | 449.1273 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction