missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Type | Tunicamycin |
| Physical Form | Powder |
| Color | Tan to White |
| Quantity | 1 mg |
| Formula Weight | 840 |
| Chemical Name or Material | Tunicamycin |
Safety and Handling
Product Identifier
- Tunicamycin
- Danger
- Acute toxicity Category 2
- H300-Fatal if swallowed.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P264-Wash thoroughly after handling.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P321-Specific treatment (see on this label).
- P330-Rinse mouth.
- P405-Store locked up.
- P501-Dispose of contents/container to .
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?