missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tunicamycin, MedChemExpress
Click to view available options
Quantity:
2 mg
5 mg
Unit Size:
2mg
5mg
Description
Tunicamycin causes accumulation of unfolded proteins in cell endoplasmic reticulum (ER) and induces ER stress, and causes blocking of DNA synthesis and cell cycle arrest in G1 phase. Tunicamycin inhibits gram-positive bacteria, yeasts, fungi, and viruses and has anti-cancer activity.Tunicamycin increases exosome release in cervical cancer cells.
Specifications
Specifications
| Chemical Name or Material | Tunicamycin |
| Physical Form | Solid |
| CAS | 11089-65-9 |
| Color | Light Yellow |
| Quantity | 2 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C39H64N4O16 |
| For Use With (Application) | COVID-19-anti-virus |
| Solubility Information | DMSO : 40 mg/mL (Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble) |
| SMILES | O=C1NC(N([C@H]2[C@H](O)[C@H](O)[C@@H](C(C[C@@H]3[C@H](O)[C@H](O)[C@@H](NC(/C=C/[(CH2) 8-11]C(C)C)=O)[C@H](O[C@H]4O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4NC(C)=O)O3)O)O2)C=C1)=O |
| Vis mere |
Research purposes only
Korrektion af produktindhold
Fisher Scientific Encompass-programmet tilbyder varer, som ikke er en del af vores distributionsportefølje. Disse produkter har typisk ikke billeder eller detaljerede beskrivelser. Vi er dog forpligtet til at forbedre din shoppingoplevelse. Brug formularen nedenfor til at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion