missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tunicamycin, MedChemExpress
Description
Tunicamycin causes accumulation of unfolded proteins in cell endoplasmic reticulum (ER) and induces ER stress, and causes blocking of DNA synthesis and cell cycle arrest in G1 phase. Tunicamycin inhibits gram-positive bacteria, yeasts, fungi, and viruses and has anti-cancer activity.Tunicamycin increases exosome release in cervical cancer cells.
Specifications
Specifications
| Chemical Name or Material | Tunicamycin |
| Physical Form | Solid |
| CAS | 11089-65-9 |
| Color | Light Yellow |
| Quantity | 5 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C39H64N4O16 |
| For Use With (Application) | COVID-19-anti-virus |
| Solubility Information | DMSO : 40 mg/mL (Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble) |
| SMILES | O=C1NC(N([C@H]2[C@H](O)[C@H](O)[C@@H](C(C[C@@H]3[C@H](O)[C@H](O)[C@@H](NC(/C=C/[(CH2) 8-11]C(C)C)=O)[C@H](O[C@H]4O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4NC(C)=O)O3)O)O2)C=C1)=O |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?