missing translation for 'onlineSavingsMsg'
Learn More

Tungstic(VI) acid

Product Code. 11354929
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11354929 100 g 100g
11364929 25 g 25g
11374929 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11354929 Supplier Thermo Scientific Alfa Aesar Supplier No. 082118.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7783-03-1 | H2O4W | 249.85 g/mol

Applied as flavoring agent and fragrance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-03-1
Molecular Formula H2O4W
Molecular Weight (g/mol) 249.85
MDL Number MFCD00011336
InChI Key CMPGARWFYBADJI-UHFFFAOYSA-L
Synonym tungstic acid, tungstic vi acid, dihydroxy dioxo tungsten, dihydrogen wolframate, wolframsaure, tunstic acid, dihydroxidodioxidotungsten, tungstic acid h2wo4, unii-j4d6k0rx2g
PubChem CID 1152
ChEBI CHEBI:36272
IUPAC Name dihydroxy(dioxo)tungsten
SMILES O[W](=O)(=O)O

Specifications

Boiling Point 1473°C
Melting Point 100°C (decomposition)
Physical Form Powder
Quantity 100 g
Solubility Information Insoluble in water and acid. Soluble in alkaline solution.
Formula Weight 249.86
Refractive Index 2.24
Density 5.5 g/mL
Chemical Name or Material Tungstic(VI) acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.