missing translation for 'onlineSavingsMsg'
Learn More

Tungstic Acid, TRC

Product Code. 30454928
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30454928 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30454928 Supplier Toronto Research Chemicals Supplier No. TRCT89727050G
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tungstic Acid
CAS 7783-03-1
Molecular Formula H2O4W
Quantity 50 g
Synonym Dihydrogen Tetraoxotungstate(2-), Tungsten Acid (H2WO4), Tungsten Hydroxide Oxide, W(OH)2O2), Tungstic Acid, Tungstic(VI) Acid, Dihydrogen Tungstate (H2WO4)
InChI Formula InChI=1S/2H2O.2O.W/h2*1H2;;;/q;;;;+2/p-2
SMILES O[W](=O)(=O)O
IUPAC Name dihydroxy(dioxo)tungsten
Molecular Weight (g/mol) 249.85
Formula Weight 249.9462
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.