missing translation for 'onlineSavingsMsg'
Learn More

Tungstic acid, 99+%

Product Code. 10107423 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10107423 500 g Plastic bottle 500g
10318300 5 g Glass bottle 5g
10134740 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10107423 Supplier Thermo Scientific Acros Supplier No. 207615000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7783-03-1 | H2O4W | 249.86 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-03-1
Molecular Formula H2O4W
Molecular Weight (g/mol) 249.86
MDL Number MFCD00011336
InChI Key CMPGARWFYBADJI-UHFFFAOYSA-L
IUPAC Name dioxotungstendiol
SMILES O[W](O)(=O)=O

Specifications

Color Green-Yellow to Yellow
Physical Form Powder
Quantity 500 g
Assay Percent Range 99% min
Linear Formula H2WO4
Merck Index 15, 9998
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in hf and alkalies
Formula Weight 249.86
Percent Purity ≥99%
Packaging Plastic bottle
Specific Gravity 5.5
Density 5.5000g/mL
Chemical Name or Material Tungstic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.